Separating funnels, pear shaped
5000 ml, 34/35 NS
Catalog # 511-0759
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Without graduation
- Very good chemical resistance
- Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.
Specifications
- Socket size:34/35 NS
- Capacity:5000 ml