Conical separating funnels
1000 ml, 29/32 NS
Catalog # 511-0763
Supplier: SciLabware
Aller à :
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.
Spécifications
- Socket size:29/32 NS
- Capacity:1000 ml
- Seq:05