Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
250 ml, 24/29 NS
Catalog # 511-0761
Supplier:   SciLabware
Conical separating funnels
Catalog # 511-0761
Supplier:  SciLabware
Conical separating funnels
Conical separating funnels
250 ml, 24/29 NS
Price
119,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.
Specifications
  • Socket size:
    24/29 NS
  • Capacity:
    250 ml
  • Seq:
    03