Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cleanroom tapes

Cleanroom tape, 50,8 mm
  817-0072
 :   ULTRATAPE
Cleanroom tapes
  817-0072
 :  ULTRATAPE
Cleanroom tapes

Cleanroom tape, 50,8 mm

 
469,00 €
   
1 * 3 ROU
 

These tapes are made of polyolefin with acrylic adhesive.

  • Ultra low outgassing
  • Low sulfur, low halogen and low heavy metal
  • Adhere lightly to plastics for easy removal
  • Leaves no residue and particulates

Designed specifically for use inside semiconductor and critical environment cleanrooms. It contains extremely low leachable metals, eliminating the concern of contamination.

Length: 54,8 m

Colour: White

 
  • Width:
    50,8 mm
  • Description:
    Cleanroom tape