Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buchner funnels

Buchner funnels

30 mm, 35 ml, P100
Catalog # 511-0801
Supplier:   DIXON SCIENTIFIC
Buchner funnels
Catalog # 511-0801
Supplier:  DIXON SCIENTIFIC
Buchner funnels
Buchner funnels

30 mm, 35 ml, P100

Price
88,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass

  • With sintered glass disc
  • Very good chemical resistance
  • High temperature resistance

For paper or membrane filtration disc.

Specifications
  • Plate Ø:
    30 mm
  • Porosity:
    P100
  • Capacity:
    35 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.