Mops, PowerHead™
PowerHead™ wide pore foam, 356 mm
Catalog # 129-0525
Supplier: MICRONOVA
Jump to:
Product Details & Documents
Mop head is constructed of polyester-covered foam mop heads are available both nonsterile and sterile for critical cleaning. Absorbent foam mop head is for pharmaceutical production and buffer areas. Wide-pore foam mop head and dense urethane foam mop head are for cleanroom support and maintenance areas.
- Stainless steel PowerHead™ handles hold and wring the various mop heads
- Autoclavable
- A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles
Ordering information: All components are ordered separately.
Specifications
- Désignation:PowerHead™ wide pore foam, 356 mm
- Seq:08