Mops, PolyGen™
Mop head
Catalog # 129-0167
Supplier: MICRONOVA
Jump to:
Product Details & Documents
The dual-action abrasive mesh is specially adapted to pharmaceutical and biotech projects. Heads used directly with the fibreglass/PP handle or attached to the universal handle via the quick disconnect adapter.
- Mop can clean to a dry finish or apply disinfectants requiring extended drying time
- Special herringbone weave ensures no raw edges to guarantee low particulation
- Low-linting polyester looped strands
- Reusable, launderable, and autoclavable
Ordering information: Mop heads and handle must be ordered separately.
Specifications
- Description:Mop head
- Cat. No.:129-0167
- Environmentally Preferable:Waste Reducing
Accessories
Product Information
- Catalog #AvailabilityPrice
- MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesMicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles129-0170Log in for availability1 * 1 ST - 137,00 €
Product Information
- Catalog #LengthMaterialAvailabilityPrice
- Stainless steel quick disconnect extendable handleStainless steel quick disconnect extendable handle129-0551965 - 1829 mmStainless steel 304Log in for availability1 * 1 ST - 314,00 €