Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test tubes without rim, medium wall

Test tubes without rim, medium wall

150 mm, 18 mm
Catalog # 212-7034
Supplier:   MURRAY S
Test tubes without rim, medium wall
Catalog # 212-7034
Supplier:  MURRAY S
Test tubes without rim, medium wall
Test tubes without rim, medium wall

150 mm, 18 mm

Price
136,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Borosilicate glass, grade 3.3, SAMCO '33'. Medium walled.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.

Certifications: ISO 4142:2002

Specifications
  • Ext.Ø:
    18 mm
  • Length:
    150 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.