Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pyrosol ® Buffer without pH indicator, 5 vials per pack, each vial contains 5.5 ml 1 * 5 items
Catalog # CAPEBC051-5
Supplier:
Associates of Cape Cod
Expand image
Pyrosol ® Buffer without pH indicator, 5 vials per pack, each vial contains 5.5 ml 1 * 5 items
Catalog # CAPEBC051-5
Supplier:
Associates of Cape Cod
Frequently Bought Together
Accessories for anaerobic systems, anaerobic incubation system, Anaerocult®
Supplier: Merck
Dip slides for surfaces and in liquids, Envirocheck®
Supplier: Merck
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Contact slides
Supplier: VWR Chemicals
VWR® Dip Slides
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Bioindicator, Sterikon® plus
Supplier: Merck
Accessories
Supplier: Merck
Dehydrated culture media
Supplier: Merck
VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
Control Standard Endotoxin (CSE) E.coli O113:H10 1 * 0,5 µG
Supplier: Associates of Cape Cod
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
VWR® Contact Plates for Surface and Air Control, Irradiated
Supplier: VWR Chemicals
Pyrotell Multi-test (approx.50 test) 5 ml/vial 0.03 EU/ml, 5 vials pack 1 * 5 items
Supplier: Associates of Cape Cod