Order Entry
Belgium
Orders LinkContactUsLinkComponent
Spoon spatulas, micro

Spoon spatulas, micro

6×8×1,5 mm, 0,2 ml
Catalog # 231-0392
Supplier:   NICKEL ELECTRO
Spoon spatulas, micro
Catalog # 231-0392
Supplier:  NICKEL ELECTRO
Spoon spatulas, micro
Spoon spatulas, micro

6×8×1,5 mm, 0,2 ml

Price
147,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

Stainless steel.

  • One end flattened, the other end spoon-formed
  • Good chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Specifications
  • Capacity:
    0,2 ml
  • Spoon size:
    6×8×1,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.