Scoops
42 ml, 120 mm, 30 mm
NICK4023
Stainless steel. Round bottom. Ideal for use as weighing boats etc.
- Excellent chemical resistance
- Excellent thermal resistance
- Smooth surfaces for fast, easy cleaning
- Largeur:30 mm
- Capacité:42 ml
- Longueur:120 mm



