Charts
C477, +40...+110 °C; 0 - 100 % RH, 7 days
Catalog # 620-0416
Supplier: DICKSON
Aller à :
Product Details & Documents
Documents
DICKC652_TDS_EN
Datasheet
Spécifications
- Description:C477
- Seq:61
- Range:+40...+110 °C; 0 - 100 % RH
- Recording time:7 days
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



