Order Entry
Belgium
Orders LinkContactUsLinkComponent
Charts

Charts

C473, –20...+50 °C; 0 - 100 % RH, 7 days
Catalog # 620-0414
Supplier:   DICKSON
Charts
Catalog # 620-0414
Supplier:  DICKSON
Charts

C473, –20...+50 °C; 0 - 100 % RH, 7 days

Price
193,00 €
Log in for availability
1 * 60 ST
Product Details & Documents

Documents

Specifications
  • Description:
    C473
  • Seq:
    13
  • Range:
    –20...+50 °C; 0 - 100 % RH
  • Recording time:
    7 days
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.