Charts
C417, –20...+120 °F; 0 - 100 % RH, 7 days
N° de catalogue 620-0412
Fournisseur: DICKSON
Jump to:
Product Details & Documents
Documents
DICKC652_TDS_EN
Datasheet
Specifications
- Description:C417
- Seq:18
- Range:–20...+120 °F; 0 - 100 % RH
- Recording time:7 days
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



