Fractionating columns, Dufton, Quickfit®
300 mm, 425 mm, 19/26 NS, 19/26 NS
201-8388
PYREX®, borosilicate glass
- Very good chemical resistance
- High temperature resistance
Comprises column with accurately fitted Dufton glass spiral. Good efficiency - liquid spiralling down column meets ascending vapour. Spiral can be removed for cleaning.
- Cone size:19/26 NS
- Socket size:19/26 NS
- Usable length:300 mm
- Seq:03
- Total length:425 mm



