Orders LinkContactUsLinkComponent
Filtration funnels, Quickfit®
Filtration funnels, Quickfit®
P40, 19/26 NS, 20 mm
Catalog # 201-8684
Supplier:   SciLabware
Filtration funnels, Quickfit®
Catalog # 201-8684
Supplier:  SciLabware
Filtration funnels, Quickfit®
Filtration funnels, Quickfit®
P40, 19/26 NS, 20 mm
Price
110,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc.
Cone connection.
Specifications
  • Cone size:
    19/26 NS
  • Plate Ø:
    20 mm
  • Porosity:
    P40