Order Entry
Belgium
Orders LinkContactUsLinkComponent
Büchner funnels, Quickfit®

Büchner funnels, Quickfit®

90 mm, 105 ml, 3, 24/29 NS
Catalog # 201-8682
Supplier:   SciLabware
Büchner funnels, Quickfit®
Catalog # 201-8682
Supplier:  SciLabware
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®

90 mm, 105 ml, 3, 24/29 NS

Price
132,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PYREX®, borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With sintered glass disc and integral ground glass cone.

Specifications
  • Cone size:
    24/29 NS
  • Plate Ø:
    90 mm
  • Porosity:
    3
  • Capacity:
    105 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.