Biopsy processing/embedding cassettes, Unisette™ M506
Pale orange
720-1575
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Integral lid
- Front face inclined at an angle of 45° so that the cassette can be used with certain types of cassette labelling devices
- Available in 11 non cytotoxic, non metallic colours and three fluorescent colours
Packaging: Each case contains three boxes of 500 cassettes each.
- Colour:Pale orange
- Seq:06
- Cat. No.:720-1575