Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Unisette™ M506
Biopsy processing/embedding cassettes, Unisette™ M506
Yellow
Catalog # 720-1573
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M506
Catalog # 720-1573
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M506
Biopsy processing/embedding cassettes, Unisette™ M506
Yellow
Price
267,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Integral lid
  • Front face inclined at an angle of 45° so that the cassette can be used with certain types of cassette labelling devices
  • Available in 11 non cytotoxic, non metallic colours and three fluorescent colours
Packaging: Each case contains three boxes of 500 cassettes each.
Specifications
  • Colour:
    Yellow
  • Seq:
    04