Tissue processing/embedding cassettes, Unisette™, M505
Purple
Catalog # 720-1550
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505
Catalog # 720-1550
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.
- Flow-through slots maximise fluid exchange and ensure effective drainage
- Resistant to histological solvents
- Integral lid
- Front writing face inclined 35°
- Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours
Packaging: Each case contains three boxes of 500 cassettes.
Specifications
- Colour:Purple
- Seq:10
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



