Tissue processing/embedding cassettes, Unisette™, M505
White
Catalog # 720-1556
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505
Catalog # 720-1556
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.
- Flow-through slots maximise fluid exchange and ensure effective drainage
- Resistant to histological solvents
- Integral lid
- Front writing face inclined 35°
- Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours
Packaging: Each case contains three boxes of 500 cassettes.
Specifications
- Colour:White
- Seq:01



