Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flasks, round bottom, Quickfit® ground joints

Flasks, round bottom, Quickfit® ground joints

150 ml, 29/32 NS, 120 mm
Catalog # 201-8482
Supplier:   SciLabware
Flasks, round bottom, Quickfit® ground joints
Catalog # 201-8482
Supplier:  SciLabware
Flasks, round bottom, Quickfit® ground joints
Flasks, round bottom, Quickfit® ground joints

150 ml, 29/32 NS, 120 mm

Price
98,90 €
Log in for availability
1 * 5 ST
Product Details & Documents

PYREX®, borosilicate glass, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With white, matt labelling area.

Certifications: ISO 1773, ISO 4797.

Documents

Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    150 ml
  • Height:
    120 mm
  • Seq:
    20
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.