Flasks, round bottom, Quickfit® ground joints
150 ml, 24/29 NS, 115 mm
Catalog # 201-8481
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certifications: ISO 1773, ISO 4797.
Documents
Specifications
- Socket size:24/29 NS
- Capacity:150 ml
- Height:115 mm
- Seq:18



