Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® I, M498

Tissue processing/embedding cassettes, Histosette® I, M498

Turquoise
Catalog # 720-1534
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M498
Catalog # 720-1534
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M498
Tissue processing/embedding cassettes, Histosette® I, M498

Turquoise

Price
229,00 €
Connexion Pour la disponibilité
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Resistant to histological solvents
  • Flow-through slots maximise fluid exchange and ensure proper drainage
  • Integral lid
  • 45° angle writing area to enable use with some automated cassette-labelling instruments
Spécifications
  • Colour:
    Turquoise
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.