Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
24/29 NS, 24/29 NS
Catalog # 201-8574
Supplier:   SciLabware
Multiple adapters, Quickfit®
Catalog # 201-8574
Supplier:  SciLabware
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 24/29 NS
Price
98,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, straight.
  • Both sockets are the same size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters with two parallel necks/sockets and a cone.
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    24/29 NS
  • Seq:
    04