Dressing jars
Lids, moulded glass, circular, for jars of O.Ø 100 mm
Catalog # MURRG501/01
Supplier: MURRAY S
Jump to:
Product Details & Documents
Dressing jars, made of moulded glass, round.
- Good chemical resistance
- Lids must be ordered separately
Specifications
- Description:Lids, moulded glass, circular, for jars of O.Ø 100 mm
- Seq:11