Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sample vials, trident vials

Tall, 7,00 ml, 17×58 mm, Yes
  212-7055
 :   MURRAY S
Sample vials, trident vials
  212-7055
 :  MURRAY S
Sample vials, trident vials
Sample vials, trident vials

Tall, 7,00 ml, 17×58 mm, Yes

 
287,00 €
   
1 * 399 ST
 

Vials made from neutral glass with PP screw cap. Designed for a wide variety of uses in the laboratory, production, more.

  • Robust and economical, will withstand moderate temperature change
  • Screw cap with foil coated insert for a better seal
  • Easy handling

 : For production related reasons, the number of vials per box may vary.

 
  • Cap included:
    Yes
  • Ø×H:
    17×58 mm
  • Capacity:
    7,00 ml
  • Description:
    Tall
  • Seq:
    03