Inoculating needles
Needle, Green, 20/bag
Supplier: MEDICAL WIRE EQUIPMENT
Jump to:
Product Details & Documents
PS, sterile. Disposable plastic inoculating needles.
- Moderately flexible
- Colour coded plastic
Documents
Specifications
- Packed:20/bag
- Colour:Green
- Description:Needle