Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flat optical windows

Flat optical windows

Flat optics ring for spring clap, Duran glass, 21 mm, 1 mm
Catalog # HELL201-1-23
Supplier:   Hellma
Flat optical windows
Catalog # HELL201-1-23
Supplier:  Hellma
Flat optical windows
Flat optical windows

Flat optics ring for spring clap, Duran glass, 21 mm, 1 mm

Price
120,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Optical glass cells and elements dedicated to special applications with detachable window.

Suitable for high precision analytical techniques.

Specifications
  • Window material:
    Duran glass
  • Description:
    Flat optics ring for spring clap
  • Dimensions:
    21 mm
  • Thickness:
    1 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.