HTS 24-multiwell insert system, Falcon®
With 24-well plate and lid, 8 µm
Catalog # 734-0383
Supplier: Corning
HTS 24-multiwell insert system, Falcon®
Catalog # 734-0383
Supplier: Corning
Jump to:
Product Details & Documents
The Falcon® HTS 24-multiwell insert system is designed to automate many commonly used membrane-based cell assays and increase the productivity and throughput of these assays in the drug discovery process. Available in a choice of membrane pore sizes (1, 3 and 8 µm), 24-multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability and absorption studies (Caco-2 cells), chemotaxis, cell migration and invasion assays.
- All the benefits of individual cell culture inserts in an automation-friendly format that is compatible with most robotic and fluid handling equipment
- Handle 24 inserts simultaneously, as all 24 inserts are part of a single unit that is compatible with Falcon® 24-well plates and feeder tray
Pore density: 1 µm insert (pores/cm²): 1,6×10⁶
Pore density: 3 µm insert (pores/cm²): 8,0×10⁵
Pore density: 8 µm insert (pores/cm²): 1,0×10⁵
Documents
Certificates
Search by Lot NumberSpecifications
- Pore size:8 µm
- Description:With 24-well plate and lid
- Cat. No.:734-0383
- Environmental certification:ISO 14001 - Bedford
- Effective growth area of membrane:0,3 cm²
- Suggested media volume in well:1000 - 4000 µl
- Insert height:18 mm
- Suggested media volume in insert:300 - 500 µl
- Effective diameter of membrane:6,5 mm
- Environmentally Preferable:Low Manufacturing Impact
- Distance from membrane to bottom of well:2,0 mm
- Effective growth area in 24-well plate:2,0 cm²
Accessories
Product Information
- Catalog #AvailabilityPrice
- Feeder tray specifically designed for use with Falcon® 24-multiwell insert systemsFeeder tray specifically designed for use with Falcon® 24-multiwell insert systems734-1325Log in for availability1 * 5 ST - 140,34 €



