HTS 24-multiwell insert system, Falcon®
Product 734-0379 is een vervanging voor 734-0378, maar verschilt per merk, verpakking of hoeveelheid.
Product Details & Documents
The Falcon® HTS 24-multiwell insert system is designed to automate many commonly used membrane-based cell assays and increase the productivity and throughput of these assays in the drug discovery process. Available in a choice of membrane pore sizes (1, 3 and 8 µm), 24-multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability and absorption studies (Caco-2 cells), chemotaxis, cell migration and invasion assays.
- All the benefits of individual cell culture inserts in an automation-friendly format that is compatible with most robotic and fluid handling equipment
- Handle 24 inserts simultaneously, as all 24 inserts are part of a single unit that is compatible with Falcon® 24-well plates and feeder tray
Pore density: 1 µm insert (pores/cm²): 1,6×10⁶
Pore density: 3 µm insert (pores/cm²): 8,0×10⁵
Pore density: 8 µm insert (pores/cm²): 1,0×10⁵
Documents
Certificaten
Zoeken op lotnummerSpecificaties
- Pore size:1 µm
- Description:With feeder tray and lid
- Cat. No.:734-0378
- Effective growth area of membrane:0,3 cm²
- Suggested media volume in well:1000 - 4000 µl
- Insert height:18 mm
- Suggested media volume in insert:300 - 500 µl
- Effective diameter of membrane:6,5 mm
- Distance from membrane to bottom of well:2,0 mm
- Effective growth area in 24-well plate:2,0 cm²
Accessories
Productinformatie
- Catalogus #DescriptionBeschikbaarheidPrice
- 734-1325Feeder tray specifically designed for use with Falcon® 24-multiwell insert systemsLog in voor beschikbaarheid1 * 5 ST - 140,34 €
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



