Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Certified colour reference standards

Pt-Co 5 mg Pt/l, certified under ISO 9001 quality system, 500 mL
Catalog # 633-0008
Supplier:   Lovibond Tintometer
Certified colour reference standards
Catalog # 633-0008
CAS Number:  
Certified colour reference standards
Certified colour reference standards

Pt-Co 5 mg Pt/l, certified under ISO 9001 quality system, 500 mL

CAS Number:  
Price
288,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

For routine calibration of colorimeters and verification of test data.

Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.

 

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Pt-Co 5 mg Pt/l, certified under ISO 9001 quality system

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.