Orders LinkContactUsLinkComponent
Column packing support, Quickfit®
Column packing support, Quickfit®
21 mm, 24/29 NS
Catalog # 201-8369
Supplier:   SciLabware
Column packing support, Quickfit®
Catalog # 201-8369
Supplier:  SciLabware
Column packing support, Quickfit®
Column packing support, Quickfit®
21 mm, 24/29 NS
Price
22,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
Designed to support Raschig rings within plain fractionating columns. Support is passed through socket of column and rests at the constriction above the cone.
Specifications
  • Cone size:
    24/29 NS
  • Ø:
    21 mm
  • Seq:
    02