Orders LinkContactUsLinkComponent
Torches
Torches
Low flow quartz torch with 2,4 mm ID injector tube
Catalog # VARI2010090400
Supplier:   VARIAN
Torches
Catalog # VARI2010090400
Supplier:  VARIAN
Torches
Torches
Low flow quartz torch with 2,4 mm ID injector tube
Price
700,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These torches light easily and stay lit, reducing downtime. With even and stable plasma to deliver reproducible results.
Specifications
  • Description:
    Low flow quartz torch with 2,4 mm ID injector tube
  • Seq:
    04
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare quartz torch bonnet for radial ICP-OES
    Spare quartz torch bonnet for radial ICP-OES
    VARI2010070790
    Log in for availability
    1 * 1 ST - 161,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Torch alignment tool for radial ICP-OES
    Torch alignment tool for radial ICP-OES
    VARI7210020700
    Log in for availability
    1 * 1 ST - 138,00 €
Product Information
  • Catalog #
    Availability
    Price
  • GazFit fittings for one-piece and semi-demountable ICP-OES torches
    GazFit fittings for one-piece and semi-demountable ICP-OES torches
    VARI7910051290
    Log in for availability
    1 * 1 ST - 221,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Transfer tube FEP for axial ICP
    Transfer tube FEP for axial ICP
    VARI7910051590
    Log in for availability
    1 * 1 ST - 221,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Transfer tube FEP Sturman-Masters spray chamber for radial ICP
    Transfer tube FEP Sturman-Masters spray chamber for radial ICP
    VARI7910051490
    Log in for availability
    1 * 1 ST - 221,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Kit Torch Fittings
    Kit Torch Fittings
    VARI9910107100
    Log in for availability
    1 * 1 ST - 98,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Pre-optics cone for Liberty Series axial ICP-OES
    Pre-optics cone for Liberty Series axial ICP-OES
    VARI3610007300
    Log in for availability
    1 * 1 ST - 818,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Esteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OES
    Esteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OES
    VARI8110002800
    Log in for availability
    1 * 1 ST - 52,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare semi-demountable quartz torch body for radial ICP-OES
    Spare semi-demountable quartz torch body for radial ICP-OES
    VARI2010072900
    Log in for availability
    1 * 1 ST - 1 060,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Quartz injector tube, 0,8 mm
    Quartz injector tube, 0,8 mm
    VARI2010077500
    Log in for availability
    1 * 1 ST - 326,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare semi-demountable quartz torch body for axial ICP-OES
    Spare semi-demountable quartz torch body for axial ICP-OES
    VARI2010083400
    Log in for availability
    1 * 1 ST - 1 060,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Quartz injector tube, 1,4 mm
    Quartz injector tube, 1,4 mm
    VARI9910057000
    Log in for availability
    1 * 1 ST - 1 070,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Injector holder kit, includes O-ring
    Injector holder kit, includes O-ring
    VARI9910057300
    Log in for availability
    1 * 1 ST - 170,00 €
Product Information
  • Catalog #
    Availability
    Price
  • O-rings kit
    O-rings kit
    VARI9910108200
    Log in for availability
    1 * 1 ST - 30,00 €