Order Entry
Belgium
Orders LinkContactUsLinkComponent
Quinoline ≥98%

Quinoline ≥98%

Glass bottle for solids, 1 kg
Catalog # 43225.A1
Toxic
Health
Irritant
Quinoline ≥98%
Catalog # 43225.A1
CAS Number:  91-22-5
Quinoline ≥98%
Quinoline ≥98%

Glass bottle for solids, 1 kg

CAS Number:  91-22-5
Price
244,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Purity: 98% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Physical Form: Liquid Density: 1.090 Refractive Index: 1.6260 Sensitivity: Light Sensitive and Hygroscopic Appearance: Colourless to pale yellow pH: 7.3 Melting Point: -15° Boiling Point: 237° Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736

  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:14,8068
  • Boiling Pt:237 °C
  • Melting Pt:–15 °C
  • Density:1.090
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications