Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 28%

Ammonia solution 28%

Glass bottle, 500 mL
Catalog # L13168.AP
Corrosive
Irritant
Environment
Ammonia solution 28%
Catalog # L13168.AP
CAS Number:  1336-21-6
Ammonia solution 28%
Ammonia solution 28%

Glass bottle, 500 mL

CAS Number:  1336-21-6
Price
27,70 €
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Odour: Ammonia-like pH: 12 Merck Reference: 14,494 MDL No.: MFCD00011418

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Merck Index:14,494
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.90
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.