Bis-trispropane ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 64431-96-5 Molecular Formula: CH2[CH2NHC(CH2OH)3]2 Formula Weight: 282.34 Physical Form: Powder Melting Point: 168-170° MDL No.: MFCD00004689
Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
- CAS Number:64431-96-5
- Formula:C₁₁H₂₆N₂O₆
- MDL Number:MFCD00004689
- Molecular Weight:282.34 g/mol
- Melting Pt:168…170 °C
- Storage Temperature:Ambient
- EINECS:264-899-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



