Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methyl red in aqueous solution

Methyl red in aqueous solution

Plastic bottle, 100 mL
Catalog # LOVI451631
Supplier:   Lovibond
Methyl red in aqueous solution
Catalog # LOVI451631
Supplier:  Lovibond
CAS Number:  493-52-7
Methyl red in aqueous solution

Plastic bottle, 100 mL

CAS Number:  493-52-7
Price
36,50 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • EINECS:207-776-1
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL

Specifications