Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Filtration funnels

20 ml, 130 mm, 75 mm, 3, 8 mm, 65 mm
  BIBB3730/06
 :   SciLabware
Filtration funnels
  BIBB3730/06
Filtration funnels
Filtration funnels

20 ml, 130 mm, 75 mm, 3, 8 mm, 65 mm

 
217,00 €
   
1 * 5 ST
 

Pyrex® borosilicate glass.

  • Sintered glass disc
  • Filter paper is not required for filtration of solutions
  • Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
 
  • Porosity:
    3
  • Capacity:
    20 ml
  • Length:
    130 mm
  • Seq:
    02
  • Int.Ø top:
    65 mm
  • Stem O-Ø:
    8 mm
  • Stem length:
    75 mm