Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
19/26 NS, 19/26 NS
Catalog # 201-3000
Supplier:   SciLabware
Multiple adapters, Quickfit®
Catalog # 201-3000
Supplier:  SciLabware
Multiple adapters, Quickfit®
19/26 NS, 19/26 NS
Price
66,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two necks. One vertical and one angled at 45°.
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    19/26 NS
  • Seq:
    01