Orders LinkContactUsLinkComponent
Ampoule breakers
Ampoule breakers
Ampoule breakers, HDPE, 5 - 10 ml glass ampoules
Catalog # AZLOAWY702
Supplier:   SciLabware
Ampoule breakers
Catalog # AZLOAWY702
Supplier:  SciLabware
Ampoule breakers
Ampoule breakers
Ampoule breakers, HDPE, 5 - 10 ml glass ampoules
Price
637,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Ampoule breakers enables safe and easy breaking of glass ampoules.
  • Broken ampoule top is retained for safe disposal
  • Choice of size to suit all common glass ampoules
Specifications
  • Description:
    Ampoule breakers, HDPE
  • Seq:
    02
  • For:
    5 - 10 ml glass ampoules