Orders LinkContactUsLinkComponent
Ampoule breakers
Ampoule breakers
Ampoule breakers, HDPE, 1 - 4 ml glass ampoules
Catalog # AZLOAWY700
Supplier:   SciLabware
Ampoule breakers
Catalog # AZLOAWY700
Supplier:  SciLabware
Ampoule breakers
Ampoule breakers
Ampoule breakers, HDPE, 1 - 4 ml glass ampoules
Price
560,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Ampoule breakers enables safe and easy breaking of glass ampoules.
  • Broken ampoule top is retained for safe disposal
  • Choice of size to suit all common glass ampoules
Specifications
  • Description:
    Ampoule breakers, HDPE
  • Seq:
    01
  • For:
    1 - 4 ml glass ampoules