Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Boxes, 96 Well

Storage Boxes, 96 Well

Storage box with ID card and lid, without tubes, non sterile, 1,2 ml in each tube, PC
Catalog # GREI975502_1
Supplier:   Greiner Bio-One
Storage Boxes, 96 Well
Catalog # GREI975502_1
Supplier:  Greiner Bio-One
Storage Boxes, 96 Well

Storage box with ID card and lid, without tubes, non sterile, 1,2 ml in each tube, PC

Price
55,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

The storage boxes are made of polycarbonate while the individual tubes are made of biologically inert polypropylene.

  • Comes with a coding card, which enables proper storage of samples
  • Temperature resistance of the polypropylene tubes from −80 °C to +121 °C provides for a broad range of applications
Specifications
  • Capacity:
    1,2 ml in each tube
  • Description:
    Storage box with ID card and lid, without tubes, non sterile
  • Material:
    PC
  • Seq:
    02
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.