Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pasteur pipettes

Pasteur pipette, graduated, 3,5 ml, 153 mm, +, 20/bag, 500/carton
  GREI612381_500
Pasteur pipettes
  GREI612381_500
Pasteur pipettes

Pasteur pipette, graduated, 3,5 ml, 153 mm, +, 20/bag, 500/carton

 
79,80 €
   
1 * 500 ST
 

These PE pipettes are suitable for the rapid transfer of liquids.

 
  • Packed:
    20/bag, 500/carton
  • Sterile:
    +
  • Capacity:
    3,5 ml
  • Description:
    Pasteur pipette, graduated
  • Length:
    153 mm
  • Seq:
    05