Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, colour coded

pH buffer solutions, colour coded

10,00, ±0,01, Blue, 20 °C, 500 mL
Catalog # REGE11005C
Supplier:   REAGECON
pH buffer solutions, colour coded
Catalog # REGE11005C
Supplier:  REAGECON
CAS Number:  
pH buffer solutions, colour coded

10,00, ±0,01, Blue, 20 °C, 500 mL

CAS Number:  
Price
37,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

Colour coded pH buffer solutions for the calibration of pH meters.

  • Can be tested at 20 or 25 °C

Certifications: Certified by Reagecon’s ISO 17025 accredited test method and NIST traceable.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    20 °C
  • Colour:
    Blue
  • pH accuracy:
    ±0,01
  • pH value:
    10,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.