Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solution, 6,80, ±0,01, 20 °C, Plastic bottle, 500 mL
Catalog # REGE10685
Supplier:   REAGECON
Buffer solutions
Catalog # REGE10685
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

Buffer solution, 6,80, ±0,01, 20 °C, Plastic bottle, 500 mL

CAS Number:  
Price
37,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    6,80

Specifications