Order Entry
Belgium
Orders LinkContactUsLinkComponent
Counting chambers

Counting chambers

Counting chamber, bright lined, Imp. Neubauer, 0,1 mm
Catalog # 630-1509
Supplier:   MARIENFELD
Counting chambers
Catalog # 630-1509
Supplier:  MARIENFELD
Counting chambers
Counting chambers

Counting chamber, bright lined, Imp. Neubauer, 0,1 mm

Price
104,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These counting chambers are suitable for counting leucocytes and erythrocytes.

  • Double net ruling
  • Two counting grids with one division
  • Different types of design available

Certifications: IVD 98/79/EC.

Delivery information: Supplied with two cover glasses of 0,4 mm thickness.

Documents

Specifications
  • Depth of chamber:
    0,1 mm
  • Type:
    Imp. Neubauer
  • Description:
    Counting chamber, bright lined
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.