Order Entry
Belgium
Orders LinkContactUsLinkComponent
Adhesion slides

Adhesion slides

Adhesion slides with 3 wells (Ø = 15 mm)
N° de catalogue MARI0901000
Fournisseur:   MARIENFELD
Adhesion slides
N° de catalogue MARI0901000
Fournisseur:  MARIENFELD
Adhesion slides
Adhesion slides

Adhesion slides with 3 wells (Ø = 15 mm)

Prix
1 240,00 €
Connexion Pour la disponibilité
1 * 50 ST
Product Details & Documents

Adhesion slides with frosted ends and 90° ground edges.

Spécifications
  • Description:
    Adhesion slides with 3 wells (Ø = 15 mm)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.