Orders LinkContactUsLinkComponent
Temperature control jackets for viscosity cups
Temperature control jackets for viscosity cups
Temperature control jacket with tripod for ISO and AFNOR cups
Catalog # SIMEVF2068
Supplier:   Industrial Physics
Temperature control jackets for viscosity cups
Catalog # SIMEVF2068
Supplier:  Industrial Physics
Temperature control jackets for viscosity cups
Temperature control jackets for viscosity cups
Temperature control jacket with tripod for ISO and AFNOR cups
Price
621,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity beaker to the required temperature with cooled and heated liquids.
  • Adjustable feet for easy leveling
  • Built-in spirit level to set jacket exactly horizontal
  • Glass plate to prevent dripping before measurement
  • Elbow-type quick release couplings
  • Titanium anodised aluminium
Delivery information: Supplied with jacket with built-in spirit level, glass plate and tube connection.
Documents
Specifications
  • Description:
    Temperature control jacket with tripod for ISO and AFNOR cups
  • Seq:
    02