Orders LinkContactUsLinkComponent
pH indicator strips, MQuant®, Supelco®
pH indicator strips, MQuant®, Supelco®
pH indicator strips for turbid solutions, 2,0 - 9,0, 1
Catalog # 1.09502.0001
Supplier:   Merck
pH indicator strips, MQuant®, Supelco®
Catalog # 1.09502.0001
Supplier:  Merck
pH indicator strips, MQuant®, Supelco®
pH indicator strips, MQuant®, Supelco®
pH indicator strips for turbid solutions, 2,0 - 9,0, 1
Price
24,00 €
Log in for availability
1 * 1 EA
Product Details & Documents
These test strips contain special indicator dyes that are covalently bound to the reagent papers which are then adhered to the plastic strips.
  • Non bleeding - do not contaminate the solution to be measured
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
 : Analyse your pH 0 to 14 test strips (1.09535) with this reference card and the free MQuant® StripScan app. The brilliant colour-fast reference colours allow for robust measurement results even under changing lighting conditions. The StripScan app allows the customer to analyse the MQuant® test strips with their smartphone for fast, convenient and precise results which can be easily graphically presented and shared.
Specifications
  • pH gradation:
    1
  • pH range:
    2,0 - 9,0
  • Description:
    pH indicator strips for turbid solutions
  • Seq:
    14
  • Cat. No.:
    1.09502.0001
  • M-Clarity quality level:
    MQ100